Wilt u opnieuw inloggen?
Introducing our New VWR Standard Tissue Culture Line!
Introducing our New VWR Standard Tissue Culture Line!
We understand the need to stretch your dollars and the need for quality. Our New VWR Collection Tissue Culture Consumables can help you do both!
These items are vacuum-gas and plasma treated to ensure consistent cell attachment, spreading and growth.
  • Manufactured in a Class 100 cleanroom environment
  • Produced with 100% USP VI crystal virgin polystyrene
  • Sterilized by gamma irradiation
  • Certified non-pyrogenic
Providing a great value for routine tissue culture!
New VWR Standard Tissue Culture Consumables
Additional VWR Collection Essentials for your Cell Culture Needs