Wilt u opnieuw inloggen?
Quality & Regulatory Compliance System To Mitigate Risks
Quality & Regulatory Compliance System To Mitigate Risks
Redundant Manufacturing Facilities
  • Certified ISO Class 7 assembly rooms
  • Validated equipment and processes
ISO 9001 Quality System
  • Compliant with GMP/EMA requirements
  • Robust and timely complaint and CAPA programmes
  • Risk-based quality management approach
Complete Regulatory Compliance Programme
  • Animal origin-free or EMA 410-compliant
  • Sterile barrier validation per ANSI/AAMI/ISO 11607
  • ISO 11137 sterility validation
  • Endotoxin USP <85> & Particulate USP <788> on lot release available
  • BPOG standardise extractables testing protocol-capable and in use