Pipette stands
Leverancier: Hecht
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HECH42590012EA 60.9 EUR
HECH42590012 HECH42590006
Pipette stands
Pipethouders
Sloping pipette stands made of plexiglas®.
  • Suitable for measuring and volumetric pipettes
  • Available for 6 and for 12 pipettes