Multi-channel pipettes, manual, with LTS™ and RFID tag, Pipet-Lite™ XLS
Leverancier: Mettler-Toledo
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Pipet-Lite™ XLS™
RAIN17011836EA 0 EUR
RAIN17011836 RAINL8-300XLS RAIN17011835 RAINL8-50XLS RAINL12-300XLS RAINL12-50XLS RAINL8-200XLS RAINL12-10XLS RAIN17011839 RAINL8-10XLS
Multi-channel pipettes, manual, with LTS™ and RFID tag, Pipet-Lite™ XLS
Pipetten Micropipetten
This ergonomic manual pipette with comfortable handle ensures light and smooth operation and significantly reduce the risk of repetitive strain injuries. Tip shaft options included low-force LTS for improved ergonomics and universal-fit.
  • Light spring forces and low-drag seals, coupled with Magnetic Assist technology, provide low plunger forces
  • Patented LTS tip and nozzle design make mounting multi-channel tips easy
  • Finger hook allows the hand to relax between pipetting cycles
  • Silicone shock absorber in the tip ejector provides extra thumb protection by cushioning tip ejection impact
  • LTS versions reduce tip ejection forces to only 0,6 kg
  • Autoclavable (shaft and tip ejector)