Tubing kit for ketone-based organic solvents
Leverancier: Agilent Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VARI9910125300EA 0 EUR
VARI9910125300 VARI9910125100 VARI9910125000
Tubing kit for ketone-based organic solvents
Slangen Chromatography Tubing
Organic samples, ketone based organic solvents (includes Marprene™ pump tubing (one each black/black and white/white), FEP transfer tube (spray chamber to torch), drain tubing, tubing for waste line, one pack of EzyFit connectors, nebulizer capillary tubing, connectors and connecting tubing).