Iris scissors, for surgery
Leverancier: S Murray & Co
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MURR1866/05EA 99.8 EUR
MURR1866/05
Iris scissors, for surgery
Scharen
Stainless steel, high grade. Straight, sharp scissors designed for the fine detail work of ophthalmic surgery.
Certificaten: Comply with the essential requirements of the Medical Devices Directive 93/42/EEC.