Pipette tips, LTS®, for Rainin pipettes
Leverancier: Mettler-Toledo
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LTS®
RAINRC-L5000EA 0 EUR
RAINRC-L5000 RAINRT-L2000S 613-4718 RAINSS-L1000 RAINSS-L10 RAINRC-L2000 RAIN30389229 RAIN30389227 RAIN30389253 RAINRC-L20MLS RAIN30389216 RAINRT-L250 613-4773 613-4756 RAINRC-L250/10 RAIN30389214 RAINRT-L2000 613-4715 RAIN30389222 RAINRC-L20ML RAIN30389246 RAINSS-L1000S RAINSR-L1200S RAIN30389220 RAIN30389244 RAIN30389224 613-4777 RAIN17007086 613-4772 RAIN30389252 RAIN17005874 RAINRT-L5000 613-4711 RAINRC-L1000/10 RAINSS-L250 RAINSR-L250S RAINRC-L300 RAIN30389249 RAINRC-L10/10 RAIN30389251 RAINRC-L10ML RAINSS-L10S RAINRT-L5000S 613-4775 RAINSS-L300 613-4757 RAINSR-L300 RAINRC-L1200 RAINSR-L1000 RAINSR-L250 613-4758 RAINSR-L10 RAINSS-L300S RAINSR-L1000S RAINRC-L10MLS RAINSR-L10S RAINSR-L300S RAINRT-L1200
Pipette tips, LTS®, for Rainin pipettes
Pipettips
These thin-wall tips are designed for use with RAININ Pipet-Lite, Pipet-Plus and EDP3-Plus pipettes with LTS LiteTouch Tip Ejection System. 1200 µl tips are specifically designed for use with RAININ E12-1200 and E8-1200 multi-channel pipettes. The unique design prevents drop formation and eliminates the need for touch-off between dispenses. The tips are available in different packaging options.
  • Virgin PP, sterile or non sterile
  • Wide orifice tips (1,5 mm) minimise shear forces on delicate samples and flow resistance for viscous samples
  • Extended length tips (102 mm long) will reach into the bottom of blocks, tall vials, flasks, 100 mm tubes etc.
  • Low retention tips feature an ultra-hydrophobic tip surface that minimises liquid retention; ideal for pipetting proteins, enzymes and DNA
  • 20 ml tips have graduation marks
Bulk
- Packaged in zip-lock bags
- Very economical
Racks with removable cover
- Sealed bottoms and overlapping lids protect tips from dust, dirt and micro-organisms
- Two-position lid allows autoclaving with the lid in vent position
- Sturdy shelf construction allows easy tip loading with single and multi-channel pipettes
Green-Pak® Individual Refills and Green-Pak® SpaceSaver® Refills
- Environmentally friendly refills for racks; less plastic waste
- Individual refills contain 96 tips in 8×12 format, sealed in a shell of Code 1 PET recyclable plastic
- Each refill is completely sealed to protect the tips; hands and fingers never contact the tips during loading
- SpaceSaver® systems are provided with either eight or ten stacked refills
- SpaceSaver® refills are packaged in a code 1 PET sleeve which functions as an automatic tip dispenser that aligns and refills an empty rack with 96 tips at a time
StableStak™ racks
- 960 tips in five stacked double-96 racks
- Strong, flat trays for even tip loading
- Ideal for multi-channel pipetting in high-production environments
- Saves time, space and plastic waste
StableRak™
- 192 tips in one double-96 rack
- Each StableRak™ contains 192 tips in one rack instead of two