Spring tubing clips, SAMCO
Leverancier: Murray
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MURRL150/01EA 351 EUR
MURRL150/01 MURRL150/02
Spring tubing clips, SAMCO
Klemmen Slangklemmen
These spring tubing clips are made from chemical and stain resistant acetal copolymer and stainless steel.
  • Operates with parallel action to provide optimum closure of rubber and flexible PVC tubing
  • Size 8 accepts tubing up to 8 mm outside diameter
  • Size 12 accepts tubing up to 12 mm outside diameter