Laboratory syringes, pistol grip, Dosys™ 170 series
Leverancier: Socorex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Dosys™
613-4809EA 152 EUR
613-4809 613-4807 SOCO172.0510 613-4808 613-4812 SOCO172.0505 613-4813 613-4811 SOCO172.0501 613-4805 613-4806 SOCO172.0502 613-4803 613-4804 613-0398
Laboratory syringes, pistol grip, Dosys™ 170 series
Spuiten Injectiespuiten, medisch
Dosys™ 174 premium: Self-refilling, automatic syringe with detachable, Luer lock valve system. Enhanced corrosion resistance.
Dosys™ 173 classic: Self-refilling, automatic syringe with integrated, Luer lock valve system. Ideal for aquous, non corrosive liquids.
Dosys™ 172 basic: Automatic syringe without valve system. Luer lock nozzle.
Leveringsinformatie: Dosys™ 174 premium package includes syringe and 1 meter silicone feed tube, sinker, PVC protection sleeve, aspiration, vent and dosing cannulas, set of spare for valve, operating instructions. Dosys™ 173 classic package includes syringe and 1 meter silicone feed tube, sinker, PVC protection sleeve, aspiration and vent cannulas, set of spare for valve, operating instructions. Dosys™ 172 basic package includes syringe, PVC protection sleeve and operating instructions.