Workstations for heavy metal sample analyses, behrotest® SMA-ARM
Leverancier: Behr
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
behrotest®
BEHRB00218413EA 7280 EUR
BEHRB00218413
Workstations for heavy metal sample analyses, behrotest® SMA-ARM
Destructiesystemen Mineralisatiewerkstations
The behrotest® SMA-ARM workstation, for simultaneous aqua regia digestion of up to six samples for heavy metals analysis, are supplied with a programmable microprocessor-controlled time/temperature controller as well as a fume manifold for drawing off the acid vapours. This results in greater convenience and functionality in routine laboratory operations.
The details of the components of the workstation are described in the table below: