HPLC columns, Syncronis™ C18
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Syncronis™
555-0206EA 4180 EUR
555-0206 HYPE97105-252130 HYPE97103-102130 HYPE97105-013001 HYPE97102-054630 HYPE97105-154030 HYPE97102-052130 HYPE97105-054630 HYPE97105-032130 HYPE97105-012101 HYPE97105-253030 HYPE97105-014001 HYPE97105-103030 HYPE97105-052130 HYPE97103-053030 HYPE97102-102130 HYPE97105-254030 HYPE97105-102130 HYPE97102-053030 HYPE97105-154630 HYPE97102-032130 HYPE97105-152130 HYPE97102-103030 HYPE97103-052130 HYPE97103-054630 HYPE97103-152130 HYPE97103-103030 HYPE97103-104630 HYPE97105-104630 HYPE97105-153030 HYPE97105-254630 HYPE97103-154630
HPLC columns, Syncronis™ C18
Kolommen voor chromatografie
C18 phase with a high carbon load for greater retention.
  • Highly pure, high surface area silica
  • Double endcapped for extra surface coverage
  • Highly inert towards basic compounds
Surface area: 320 m²/g
Carbon load: 16%
pH range: 2 - 9