Burettes, class B, Schellbach
Leverancier: VITLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VITL105599EA 148 EUR
VITL105599 VITL105699
Burettes, class B, Schellbach
Plastic-coated burette made of borosilicate glass 3.3, tolerances corresponding to DIN ISO 384, Class B.
  • Schellbach stripes (blue/white) and high-contrast black marking
  • PMP/PTFE stopcock turns easily and enables fine titration
  • Temperature-stable plastic coating on the tube provides splinter protection
Calibrated to deliver 'Ex'.