PCR plates, 96-well, segmented, semi-skirted, barcoded
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ABgene®
ADVABC-0900/WEA 178 EUR
ADVABC-0900/W ADVABC-0900
PCR plates, 96-well, segmented, semi-skirted, barcoded
Microplaten PCR-platen
Proprietary segmented plate design allows plates to be cut into 24- and 48-well sections. These plates are made from PP.
  • White plates are optimized for qPCR by increasing sensitivity and reducing variability in fluorescence detection
  • Cut corner: H1
  • Raised rim design around each well enables secure sealing and safeguards against evaporation
  • Uniform, thin well walls deliver maximum and consistent heat transfer
  • Semi-skirt adds rigidity and allows for labeling or barcoding
The barcodes are the industry-standard code 128. The label itself can withstand temperatures from -196 up to 120 °C to ensure that the barcodes stay on the plate throughout your entire workflow. It also has a special coating to virtually eliminate any "rubbing off" of the code during handling. The dual barcodes are located on the opposite numeric and opposite alpha sides of the plate.