Wash head for wash bottles
Leverancier: VITLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VITL83301EA 31.8 EUR
VITL83301 VITL83302 VITL83300 VITL83304 VITL83303
Wash head for wash bottles
Sluitingen Flesdoppen
Made of PP, translucent.
  • Very good chemical resistance
  • Very good temperature resistance
Wash bottle screw caps and spray tube with drawn-out tip, Precise spray jet and optimised medium backflow through the continuous and finely drawn tip of the spray tube.