Capillary columns, FS-CW 20 M-AM
Leverancier: Macherey-Nagel
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MANA733111.10EA 407 EUR
MANA733111.10 MANA733442.50 MANA733299.50 MANA733110.50 MANA733551.25 MANA733299.25 MANA733110.25
Capillary columns, FS-CW 20 M-AM
Kolommen voor chromatografie
Special polar GC phase for the analysis of amines.
  • Deactivated polyethylene glycol for aliphatic and aromatic amines
  • Max. temperature for isothermal operation 220 °C
  • Max. temperature for short isotherms in a temperature program 240 °C
Basic polyethylene glycol (20 kD) phase.
USP code: G16.