Wash bottles, narrow neck
Leverancier: VITLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VITL108792EA 0 EUR
VITL108792
Wash bottles, narrow neck
Flessen Spuitflessen
PFA, translucent, screw cap made from ETFE, spray tube from FEP.
  • Excellent chemical resistance
  • Temperature resistance -200 °C to +260 °C
  • Can be used for less critical applications for which pure PFA is not necessary
With recycled PFA content. Thus, reasonably priced and environmentally friendly. Precise spray jet and optimised medium backflow through the continuous and finely drawn tip of the spray tube.