TLC plates, silica gel 60, unmodified layers, with concentrating zone, SILGUR
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ALUGRAM®
MANA818423EA 136 EUR
MANA818423 552-0216 MANA818413 552-0214 MANA818412 MANA810123 MANA818422 552-0213 552-0215
TLC plates, silica gel 60, unmodified layers, with concentrating zone, SILGUR
TLC-platen
Unmodified standard silica layers with concentrating zone for TLC.
  • High-quality silica gel with kieselguhr zone
  • Rapid sample application
  • Always regular substance spots
Also after arbitrary application of the substances in the concentrating zone the developed chromatogram shows regular spots for a reliable detection.
Silica 60: Specific surface (BET) ~ 500 m²/g; specific pore volume 0,75 ml/g; particle size 5 – 17 μm.
Kieselguhr: Small surface of low activity.