HPTLC plates, silica gel 60, modified layers, RP-18 W/UV₂₅₄
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ALUGRAM®
552-0305EA 258 EUR
552-0305 552-0308 552-0245 552-1293 552-0304 552-0244 552-0306 552-0307 552-0246 552-0247
HPTLC plates, silica gel 60, modified layers, RP-18 W/UV₂₅₄
TLC-platen
Octadecyl-modified HPTLC silica layers available as glass plates or ALUGRAM® aluminium sheets. Recommended for aminophenol, barbiturates, preservatives, nucleobases, polycyclic aromatic, hydrocarbons, steroids, tetracyclines, and plasticisers (phthalates).
Base material: Nano-silica 60: Specific surface (BET) ~ 500 m²/g; specific pore volume 0,75 ml/g; particle size 2 – 10 μm; pH stability 2 – 10
Silica 60 for preparative plates (1000 µm): Specific surface (BET) ~ 500 m²/g; specific pore volume 0,75 ml/g; particle size 5 – 17 μm; pH stability 2 – 10
Modification: Partial octadecyl, wettable with water, carbon content 7,5%.
Indicator: Acid-resistant product with a pale blue fluorescence for short-wave UV (254 nm) and UV-absorbing substances appear as dark-blue to black spots on a light-blue background.