Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')dysprosium(III)
Leverancier: Wako
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WAKO321-86183EA 0 EUR
WAKO321-86183
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')dysprosium(III)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')dysprosium(III)
Formule: C₃₃H₅₇DyO₆
MW: 712,302 g/mol
MDL Nummer: MFCD00000017
CAS nummer: 15522-69-7
EINECS: 239-563-4