Nitrite-Nitrogen standard solution, 1000 mg/L NO₂- (as N) in Water standard for ion chromatography
Leverancier: CPAchem
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ACSDH043.W.L5EA 143 EUR
ACSDH043.W.L5 ACSDH043.W.L25 ACSDH043.W.L1
Nitrite-Nitrogen standard solution, 1000 mg/L NO₂- (as N) in Water standard for ion chromatography
Standaarden Anionische standaarden Nitriet standaarden
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022