3,5-Dimethyl-1H-pyrazole-1-carboxamide
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MAYBCD09590.250EA 0 EUR
MAYBCD09590.250 MAYBCD09590.1
3,5-Dimethyl-1H-pyrazole-1-carboxamide
3,5-Dimethyl-1H-pyrazole-1-carboxamide
Formule: C₆H₉N₃O
MW: 139,16 g/mol
MDL Nummer: MFCD00003126
CAS nummer: 934-48-5
EINECS: 213-283-2