1,2:7,8-Diepoxyoctane ≥97.0%
Leverancier: TCI EUROPE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
TCIAD1586-5GEA 55 EUR
TCIAD1586-5G TCIAD1586-25G
1,2:7,8-Diepoxyoctane ≥97.0%
1,2:7,8-Diepoxyoctane
Formule: C₈H₁₄O₂
MW: 142,20 g/mol
Kookpunt: 101°C
Vlampunt: 98°C
MDL Nummer: MFCD00005155
CAS nummer: 2426-07-5
EINECS: 219-375-9
UN: 2810
ADR: 6.1,III
https://nl.vwr.com/stibo/web/std.lang.all/83/87/28978387.jpg
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %