Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?

4'-Aminoacetanilide hydrochloride ≥98%

Leverancier: Alfa Aesar

Purity: 98%
CAS No.: 43036-07-3
Molecular Formula: C8H10N2O.HCl
Formula Weight: 186.64
Melting Point: 309-312°
Beilstein Reference: 3703679


Formule: C₈H₁₁ClN₂O
MDL Nummer: MFCD00050695
CAS nummer: 43036-07-3
EINECS: 256-052-1


 

Order

Expand All / Collapse All
Pack type VPE VWR catalogus nummer Hoeveelheid
Glass bottle for solids 100 g A13937.22
Glass bottle for solids 25 g A13937.14
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Specificaties

Melting point 309-312°
Formulegewicht 186.64
CAS No. 43036-07-3
Zuiverheid 98%
Molecular formula C8H10N2O.HCl
MDL No. MFCD00050695
Beilstein reference 3703679

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'X' if unknown for VWR Chemicals. Enter a space if unknown for Merck Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Of
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Omschrijving Catalogus nummer Availability Unit Uw prijs Price Per Hoeveelheid