Isopropyldiphenylphosphine ≥97.0% (by GC, titration analysis)
Leverancier: TCI EUROPE
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
TCIAI0583-1GEA
55
EUR
TCIAI0583-1G
TCIAI0583-5G
Isopropyldiphenylphosphine ≥97.0% (by GC, titration analysis)
Isopropyldiphenylphosphine
Formule:
C₁₅H₁₇P MW: 228,27 g/mol |
MDL Nummer:
MFCD00015028 CAS nummer: 6372-40-3 EINECS: 228-902-1 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Iodometric Titration) | min. 97.0 % |
Melting point | 39.0 to 43.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...