Glycidyl isopropyl ether for synthesis
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
M-Clarity™ quality level = MQ200
8.18598.2500EA 0 EUR
8.18598.2500 8.18598.0100 8.18598.9999
Glycidyl isopropyl ether for synthesis
Glycidyl isopropyl ether
Formule: C₆H₁₂O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005139
CAS nummer: 4016-14-2
EINECS: 223-672-9
UN: 1992
ADR: 3 (6.1),III
Order Now
Specification Test Results
Solubility 188 g/l (19 °C)
Molar Mass 116.15 g/mol
Boiling Point 137 °C (1013 hPa)
Vapor Pressure 13 hPa (25 °C)
Flash Point 33 °C
Refractive Index 1.4101 (20 °C, 589 nm)
Density 0.92 g/cm3 (20 °C)
pH 7 (188 g/l, H2O, 20 °C)