Netherlands
Order Entry
Bromate standard solution, 1,000 mg/L BrO3- in Water standard for ion chromatography
Bromate standard solution, 1,000 mg/L BrO3- in Water standard for ion chromatography
Supplier:  CPACHEM
Certificates
Bromate standard solution, 1,000 mg/L BrO3- in Water standard for ion chromatography
Supplier:  CPACHEM
Certificates
Related Information
Product Information
  • Catalog #
    Supplier #
    Concentration
    Contents
    Pack type
    Pk
    Availability
    Price
  • Bromate standard solution
    Bromate standard solution
    ACSDH020.W.L25
    H020.W.L25
    1000 mg/l
    KBrO₃ in H₂O
    Plastic bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 133,00 €
    133,00 €
    1 * 250 mL
       133,00 €
    Supplier Article Number: H020.W.L25
    Local Article Number: ACSDH020.W.L25
  • Bromate standard solution
    Bromate standard solution
    ACSDH020.W.L5
    H020.W.L5
    1000 mg/l
    KBrO₃ in H₂O
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 187,00 €
    187,00 €
    1 * 500 mL
       187,00 €
    Supplier Article Number: H020.W.L5
    Local Article Number: ACSDH020.W.L5
  • Bromate standard solution
    Bromate standard solution
    ACSDH020.W.L1
    H020.W.L1
    1000 mg/l
    KBrO₃ in H₂O
    Plastic bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL
    Supplier Article Number: H020.W.L1
    Local Article Number: ACSDH020.W.L1