Reusable earplugs, Neutron™
Leverancier: Howard Leight by Honeywell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Neutron™
111-1443EA 80.55 EUR
DALL1029810
Reusable earplugs, Neutron™
These dual-flange earplugs enable users to hear more clearly in moderate noise levels without distorted speech sounds. Thanks to the optimal attenuation across all frequencies workers stay connected to their environment. They can clearly hear voices, signals, and alarms without having to remove the earplugs.
  • Reduce hazardous noise levels without distorting the incoming sound
  • Sleek earbud design and long, streamlined flanges
  • Intuitive fit and comfortable seal inside the ear canal
  • Yellow polycord improves visibility
SNR: 20 dB
Certificaten: EN 352-2
Opgelet: * One size fits all.