Glass wool
Leverancier: Macherey-Nagel
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MANA481110EA 52.7 EUR
MANA481110 MANA481100 MANA281130 MANA281120
Glass wool
Glaswol en katoen
Filter aids transform difficult precipitates and colloidal particles into a form which can be easily filtered. When slimy and strongly lyophilic, swelling precipitates are involved, the fibres of the filter flocs prevent formation of a continuous, impermeable layer on the filter. The resulting filter cake remains porous and permeable, and clogging of the filter is prevented.