Netherlands
Order Entry
Handheld airborne particle counters, MET One HHPC
Handheld airborne particle counters, MET One HHPC
undefined
Handheld airborne particle counters, MET One HHPC
  • Catalog No:
  • 710-0979
  • 710-0982
  • 710-0980
  • Model:
  • MET One HHPC 3+, 3 channels (0,3 - 10 µm)
  • MET One HHPC 6+, 6 channels (0,3 - 10 µm)
  • MET One HHPC 2+, 2 channels (0,5 - 5 µm)
  • Flow rate:
  • 2,83 L/min, 0,3 µm/min
  • 2,83 L/min, 0,3 µm/min
  • 2,83 L/min, 0,5 µm/min
  • Material:
  • Polycarbonate
  • Polycarbonate
  • Polycarbonate
  • Power:
  • 110/240 V
  • 110/240 V
  • 110/240 V
  •  
    Pk
  • MET One HHPC 2+, 2 channels (0,5 - 5 µm)
    MET One HHPC 2+, 2 channels (0,5 - 5 µm)
    710-0980
    2089310-03
    1
    1 * 1 ST
    1 * 1 ST
    Accessories
    Specifications
    Model
    MET One HHPC 2+, 2 channels (0,5 - 5 µm)
    Environmentally Preferable
    Flow rate
    2,83 L/min, 0,5 µm/min
    Material
    Polycarbonate
    Power
    110/240 V
  • MET One HHPC 3+, 3 channels (0,3 - 10 µm)
    MET One HHPC 3+, 3 channels (0,3 - 10 µm)
    710-0979
    2089310-02
    1
    1 * 1 ST
    1 * 1 ST
    Accessories
    Specifications
    Model
    MET One HHPC 3+, 3 channels (0,3 - 10 µm)
    Environmentally Preferable
    Flow rate
    2,83 L/min, 0,3 µm/min
    Material
    Polycarbonate
    Power
    110/240 V
  • MET One HHPC 6+, 6 channels (0,3 - 10 µm)
    MET One HHPC 6+, 6 channels (0,3 - 10 µm)
    710-0982
    2089311-01
    1
    1 * 1 ST
    1 * 1 ST
    Accessories
    Specifications
    Model
    MET One HHPC 6+, 6 channels (0,3 - 10 µm)
    Environmentally Preferable
    Flow rate
    2,83 L/min, 0,3 µm/min
    Material
    Polycarbonate
    Power
    110/240 V
This product has been blocked by your organization. Please contact your purchasing department for more information