(R)-(+)-N-Benzyl-α-methylbenzylamine ≥99%, ee 96%, ChiPros®
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
L19298.06EA
73.8
EUR
L19298.06
(R)-(+)-N-Benzyl-α-methylbenzylamine ≥99%, ee 96%, ChiPros®
(R)-(+)-N-Benzyl-α-methylbenzylamine
Formule:
C₁₅H₁₇N MW: 211,31 g/mol Kookpunt: 171 °C (15 mmHg) Dichtheid: 1,010 Vlampunt: >110 °C (230 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00015010 CAS nummer: 38235-77-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...