4-Hydroxy-3-methoxybenzylamine hydrochloride for synthesis
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
M-Clarity™ quality level = MQ200
8.41494.0001EA 40.7 EUR
8.41494.0001
4-Hydroxy-3-methoxybenzylamine hydrochloride for synthesis
4-Hydroxy-3-methoxybenzylamine hydrochloride
Formule: C₈H₁₂ClNO₂
MW: 189,64 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012864
CAS nummer: 7149-10-2
EINECS: 230-468-3
Order Now
Specification Test Results
Melting Point 219 - 220 °C (decomposition)
Molar Mass 189.64 g/mol