The Leica HI1210 is a water bath with a surface that provides high thermal conductivity rates and outstanding scratch resistance due to its special plastic coating. Temperatures can be selected between ambient and 75 °C.
- LED display
- Less sensitive to water and easily cleaned
- Better contrast
- Visual indication
To increase safety and reliability the Leica HI1210 features an overheating protection system along with a stand-by mode. The broad, oversized rim of the water bath allows convenient storage for microscope slides, and the rounded inner corners of the instrument allow it to be cleaned easily and efficiently. The selected temperature set value is memorised via a built-in battery backup system. Jet black surface with scratch-proof plastic coating provides better contrast to identify sections and an easy to clean surface. Capacity: 2 L.
W×D×H: 308×340×95 mm.
Leveringsinformatie: Basic instrument, 1 dust cover , power cord "D" , set of replacement fuses 2×T 2,5 A, 1 instruction manual Leica HI1210, 1 CD including available languages.