Flattening table for clinical histopathology, Leica HI1220
Leverancier: Leica
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
720-0114EA 2050 EUR
720-0114
Flattening table for clinical histopathology, Leica HI1220
Verwarmers/drogers voor glaasjes
The Leica HI1220 flattening table has a large jet black aluminum work surface to provide high thermal conductivity rates and outstanding resistance to mechanical manipulations. Temperatures ranging from ambient to 75 °C can be selected, with work surface temperatures above 44 °C being indicated by a flashing LED, to guard against overheating of specimens. The programmed, as well as the current, temperatures are displayed on the instrument and the set values are memorized via a built-in battery backup system.
  • LED display
  • Less sensitive to water and easily cleaned
  • Better contrast
WxDxH: 308×340×95 mm
Leveringsinformatie: 1 Basic instrument, 1 dust cover , 1 power cord "D" , set of replacement fuses 2×T 2,5 A,1 instruction manual Leica HI1220, 1 CD including available languages.