VWR® RIPA Lysis Buffer, pH 7,5
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR®
N653-100MLEA 115 EUR
N653-100ML
VWR® RIPA Lysis Buffer, pH 7,5
Buffers Extractiebuffers
Lysis buffer for cultured mammalian cells contains SDS and deoxycholate for solubilisation of proteins from cytoplasm, membranes, and nuclei. Compatible with reporter assays, protein assays, immunoassays, and protein purification.
RIPA buffer contains the non ionic detergent, NP-40 Substitute, plus two ionic detergents, sodium deoxycholate and SDS.