Netherlands
Order Entry
Cation exchange chromatography columns, HiTrap™ CM FF
Cation exchange chromatography columns, HiTrap™ CM FF
Supplier:  Cytiva
Certificates
undefined
Cation exchange chromatography columns, HiTrap™ CM FF
Supplier:  Cytiva
Specifications
  • Catalog No:
  • 17-5056-01
  • 17-5155-01
  • Max. pressure:
  • 5 bar
  • 5 bar
  • pH range:
  • 4 - 13
  • 4 - 13
Specifications
Product Information
  • Catalog #
    Supplier #
    Column hardware
    Particle Size
    Bed dimensions
    Capacity
    Flow rate
    Availability
    Price
  • HiTrap™ CM FF
    HiTrap™ CM FF
    17-5056-01
    17-5056-01
    PP
    90 µm
    7×25 mm
    1 ml
    <4 ml/min
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 174,00 €
    174,00 €
    1 * 5 ST
       174,00 €
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Max. pressure
    5 bar
    pH range
    4 - 13
  • HiTrap™ CM FF
    HiTrap™ CM FF
    17-5155-01
    17-5155-01
    PP
    90 µm
    16×25 mm
    5 ml
    <20 ml/min
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 378,00 €
    378,00 €
    1 * 5 ST
       378,00 €
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Max. pressure
    5 bar
    pH range
    4 - 13