PhastTransfer™ kit
Leverancier: Cytiva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
18-1003-18EA 93.5 EUR
GEHE18-1003-18
PhastTransfer™ kit
PhastSystem™ consists of a separation-control unit for system control and electrophoresis, and a development unit for gel staining.
  • The optional PhastTransfer™ kit quickly converts the system into a semi-dry blotting unit
Electrophoresis is performed on a thermostatic plate capable of maintaining temperatures accurately from 0 - 70 °C. Multistep programming capability allows gels to be prerun, loaded and run automatically. Controlled parameters include temperature, voltage, current, power and volt hours.
Leveringsinformatie: Contains gel-backing remover (mounting block plus cutting frame), electrode cassette, blotting paper and timer.