Blocking buffer, PBS with Tween® 20, powder blend, pH 7,4
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M235-12.5G-5PKEA 158 EUR
VWRCM235-12.5G-5PK
Blocking buffer, PBS with Tween® 20, powder blend, pH 7,4
Powdered blocking buffer blend supplied in five convenient premeasured packs. Each pack prepares 1 litre of 1X solution.