Disposal Bins
Leverancier: asecos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MSGA18425EA 188 EUR
MSGA18425 MSGA18429
Disposal Bins
Bakken Afvalbakken
Sheet steel.
  • Reduce escaping vapours to a minimum
  • Safe installation directly at the workplace
  • Self-closing lid can be opened by foot pedal or by hand
  • Bottom edge with ventilation openings prevents spontaneous combustion of contents
Ideal for safe disposal of rags impregnated with solvents or other waste.