Disposal Bins
Leverancier: asecos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MSGA8640EA 459 EUR
MSGA8640 MSGA8641
Disposal Bins
Bakken Afvalbakken
Made of stainless steel.
  • Very good chemical resistance
  • Easy to clean
Collection container with lid and foot pedal. Safe disposal of rags impregnated with solvents or other waste.