Granule carriers
Leverancier: asecos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MSGA14532EA 373 EUR
MSGA14532
Granule carriers
Transportwagens
Trolley-type granule carrier for transport and storage of unused and collection for disposal of used granule.
  • Robust construction
  • Two removable plastic sumps
  • Easy handling with two guide rollers and fixed castors
Bestelinformatie: Shovels are not included and should be ordered separately.