Rapid-hyb buffer
Leverancier: Cytiva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
RPN1636EA 555 EUR
RPN1636
Rapid-hyb buffer
Buffers Buffers voor elektroforese Hybridisatiebuffers
A major limitation of membrane hybridisation is speed. At commonly used probe concentrations (2 to 10 ng/ml), hybridisation normally takes 16 hours to detect single-copy gene sequences in mammalian DNA. Use of Rapid-hyb buffer reduces this time to 2 hours using similar probe concentrations.
Store at ambient temperature. Under these conditions the product is stable for at least 3 months.