Safety blood lancets with sterile needle
Leverancier: Marienfeld
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MARI7300805EA 19.5 EUR
MARI7300805
Safety blood lancets with sterile needle
Safety lancets with gamma-sterilised needle, which remains fully secured in its casing – before and after the penetration.
  • For secured single use
  • Prevents injuries or infections and allows a safe handling
  • Different colours identify the gauge size and penetration depth
The painless penetration is effected at high speed by pushing a button.
Certificaten: Approved for in vitro diagnostic (IVD) applications according to IVD directive 98/79/EC, with CE - mark, recommended best before date and batch number for comprehensive information and traceability.