Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?
Pipette canisters, round bottom
Leverancier: BELLCO GLASS INC.
Pipette canister made of borosilicate glass, class A, type I.
  • Manufactured from Type 1, Class A borosilicate glass
  • Components will withstand repeated dry or steam heat sterilisation
Order
Expand All / Collapse All
Omschrijving VPE VWR catalogus nummer Hoeveelheid
Pipette canister, round bottom, H×Ø: 430×38 mm 6 BELC1287-00430
Pipette canister, round bottom, with stainless steel closure, for Shortie pipettes, H×Ø: 250×38 mm 6 BELC1287-38250
Pipette canister, round bottom, with stainless steel closure, H×Ø: 430×38 mm 6 BELC1287-38430
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Accessoires
Expand All / Collapse All
Accessories
Omschrijving VPE VWR catalogus nummer Hoeveelheid
Mounting plate, 90×90 mm, for use with 38mm OD glass pipette canisters 6 BELC1288-11111
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
WRITE A REVIEW
Need help?
  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.
VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.
Please confirm the industry you work in
Example: Pharmaceutical
Please confirm your role
Example: Clinical Trial Assistant
Please confirm your location
Example: New York, NY
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'X' if unknown for VWR Chemicals. Enter a space if unknown for Merck Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Of
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Omschrijving Catalogus nummer Availability Unit Uw prijs Price Per Hoeveelheid
Een nieuwe uitstraling en een hernieuwde belofte
Gebouwd op de kenmerken & middelen die u kent, geoptimaliseerd met de innovatie van Avantor ®. Ons nieuwe merk symboliseert flexibiliteit en ons vermogen om ons aan de behoeften van de klant aan te passen.
KOM MEER TE WETEN BEGIN MET WINKELEN