Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?
Desiccators, vacuum
Leverancier: SciLabware
Leveringsinformatie: Supplied wth plate.
login om uw prijzen en beschikbaarheid te zien
  Onthoud mij op dit apparaat
Expand All / Collapse All
With stopcock
Type Nominale afmeting Bruikbare hoogte Ø uitw.×h Omschrijving VPE VWR catalogus nummer Hoeveelheid
With stopcock 200 DN 178 mm 260×262 mm Vacuum desiccator, with ceramic plate 1 AZLODWA215
With stopcock 250 DN 195 mm 290×296 mm Vacuum desiccator, with ceramic plate 1 AZLODWA245
Type Diameter VPE VWR catalogus nummer Hoeveelheid
Ceramic 205 mm 1 AZLODWA205
Ceramic 236 mm 1 AZLODWA236
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Expand All / Collapse All
Type Voor VPE VWR catalogus nummer Hoeveelheid
Spare stopcock DWA215/245 desiccators 1 AZLODWA510
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Need help?
  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.
VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.
Please confirm the industry you work in
Example: Pharmaceutical
Please confirm your role
Example: Clinical Trial Assistant
Please confirm your location
Example: New York, NY
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'X' if unknown for VWR Chemicals. Enter a space if unknown for Merck Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Omschrijving Catalogus nummer Availability Unit Uw prijs Price Per Hoeveelheid