Brass (alloy 260), spheres, Ø 3.18 mm (0.125 in)
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
45139.KNEA 26.3 EUR
45139.KN
Brass (alloy 260), spheres, Ø 3.18 mm (0.125 in)
Brass (alloy 260)