Theophyllin
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
28614.237EA
0
EUR
28614.237
Theophyllin
Theophyllin
Formule:
C₇H₈N₄O₂ MW: 180,16 g/mol Smeltpunt: 274 °C Dichtheid: 1,35 g/cm³ (25 °C) |
MDL Nummer:
MFCD00079619 CAS nummer: 58-55-9 EINECS: 200-385-7 UN: 2811 ADR: 6.1,III |
Specification Test Results
Identification | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...