Acepromazine maleate
Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PROOBP001EA 0 EUR
PROOBP001
Acepromazine maleate
Acepromazine maleate
Formule: C₂₃H₂₆N₂O₅S
CAS nummer: 3598-37-6
EINECS: 222-748-9