Diphenylphosphoryl azide 98%
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
ACRO409040250EA 83.2 EUR
ACRO409040250 ACRO409045000 ACRO409041000
Diphenylphosphoryl azide 98%
Diphenylphosphoryl azide
Formule: C₁₂H₁₀N₃O₃P
MW: 275,20 g/mol
Kookpunt: 157 °C (,2 mmHg)
Dichtheid: 1,273 g/cm³
Vlampunt: 112 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00001987
CAS nummer: 26386-88-9
EINECS: 247-644-0
UN: 3278
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Clear yellow to amber Liquid
Infrared spectrum Conforms
Refractive index 1.5500 to 1.5520 (20°C, 589 nm)
GC ≥97.5 %