Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?
  • Criteria
    • In Stock (32)
  • Leverancier
  • Refine by Suppliers
    Sort by:
  • Zoek binnen resultaten
DNA/RNA reinigingskits
https://nl.vwr.com/stibo/smallweb/std.lang.all/50/96/18945096.jpg
DNA and RNA may need to be concentrated and cleaned up to remove enzymes, oligonucleotides, buffers, or chemical inhibitors. With the DNA/RNA cleanup kits available from VWR, high-quality DNA and RNA can be purified and concentrated, even from small sample volumes. These efficient and time-saving purification tools result in DNA and RNA which may be used in any common downstream application like restriction enzyme digestion, labeling, hybridisation, ligation, cloning, and sequencing.
SearchPresentationType-VERTICAL
SearchResultCount:"29"
  List view Grid view Easy View (new)
Sorteer op:
Catalogus nummer: (1B1414-200RXN)
Leverancier:  VWR Chemicals
Omschrijving:   Recombinant M-MLV Reverse Transcriptase purified from E. coli expressing the pol gene of Moloney Murine Leukemia Virus.
UOM:  1 * 1 EA
Leverancier:  GE Healthcare - Life Sciences
Omschrijving:   Voor de zuivering van DNA van kleine moleculen d.m.v. gel filtratie.
Leverancier:  GE Healthcare - Life Sciences
Omschrijving:   DNA/RNA reinigingskits, illustra™ GFX™, illustra™ GFX™ PCR DNA and Gel Band Purification Kit, 250 purifications
Leverancier:  OMEGA BIOTEK
Omschrijving:   Gelzuivering van DNA is een techniek die vaak wordt gebruikt voor het isoleren van specifieke DNA-fragmenten. De meeste methoden slagen er echter niet...
Leverancier:  OMEGA BIOTEK
Omschrijving:   E.Z.N.A.® Cycle Pure-kits zijn gemaakt voor de snelle opzuivering van enkel- of dubbelstrengs DNA na PCR of andere enzymatische reacties. De zuivering...
Catalogus nummer: (27-5325-01)
Leverancier:  GE Healthcare - Life Sciences
Omschrijving:   For rapid buffer exchange/desalting of PCR products and other DNAs in a volume of 10 to 100 μl using spin column chromatography.
UOM:  1 * 50 ST
Catalogus nummer: (17-0574-02)
Leverancier:  GE Healthcare - Life Sciences
Omschrijving:   For purification of DNA from small molecules by gel filtration.
UOM:  1 * 100 g
Leverancier:  GE Healthcare - Life Sciences
Omschrijving:   DNA/RNA reinigingskits, illustra™ MicroSpin™, illustra™ MicroSpin™ G-50 columns
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Dit artikel is vervallen en leverbaar zolang de voorraad strekt. Binnen de VWR catalogus kunt u zoeken op alternatieven. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 8  Of 29
  1  2  3  4  Volgende