Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?
  • Product category
  • Criteria
  • Leverancier
  • Refine by Suppliers
    Sort by:
  • Zoek binnen resultaten
Zuiveringskits voor nucleïnezuren
https://nl.vwr.com/stibo/smallweb/std.lang.all/96/96/18989696.jpg
SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-VERTICAL
SearchResultCount:"185"
  List view Grid view Easy View (new)
Sorteer op:
Catalogus nummer: (1.01193.0001)
Leverancier:  Merck
Omschrijving:   Zuivering van nucleïnezuren, Genomisch DNA-zuiveringskit, MagPrep®, Silica particles, MagPrep®
UOM:  1 * 1 ml
Leverancier:  VWR Chemicals
Omschrijving:   Hi-Bind® DNA mini columns are silica glass fibre columns that can bind up to 100 μg of genomic DNA or 35 μg of plasmid DNA per prep. Columns feature a...
Leverancier:  VWR Chemicals
Omschrijving:   The E.Z.N.A.® Plasmid Mini Kit I is designed to isolate up to 30 µg of high-quality plasmid DNA from 1 to 5 ml bacterial cultures in 30 minutes o...
Leverancier:  VWR Chemicals
Omschrijving:   VWR® ExoCleanUp PCR reagent is a one-step PCR clean up reagent for optimal sequencing results consisting of a balanced combination of a heat labile ex...
Leverancier:  VWR Chemicals
Omschrijving:   The Yeast Genomic DNA Purification Kit yields high molecular weight DNA from yeast using a non-enzymatic lysis buffer and no glass beads. The protocol...
Catalogus nummer: (N788-KIT)
Leverancier:  VWR Chemicals
Omschrijving:   The Phenol-free Total RNA Purification Kit provides a rapid method for the isolation and purification of total RNA from cultured animal cells, tissue ...
UOM:  1 * 1 EA
Catalogus nummer: (N643-KIT)
Leverancier:  VWR Chemicals
Omschrijving:   Zuivering van nucleïnezuren, RNA-zuiveringskit, RIBOZOL™, RiboZol™ Plus RNA Purification Kit, 2×30 ml
UOM:  1 * 1 EA
Catalogus nummer: (1B1558-KIT)
Leverancier:  VWR Chemicals
Omschrijving:   Fluorescent DNA Quantitation Kit uses fluorescent bisBenzimide H33258 dye to bind to AT sequences of the minor groove of double-stranded DNA. In the a...
UOM:  1 * 1 EA
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Dit artikel is vervallen en leverbaar zolang de voorraad strekt. Binnen de VWR catalogus kunt u zoeken op alternatieven. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 8  Of 185
  1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  Volgende